missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Thiophene-2-glyoxylate, TRC

Product Code. 30383011
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383011 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383011 Supplier Toronto Research Chemicals Supplier No. TRCE92701025G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Thiophene-2-glyoxylate
CAS 4075-58-5
Molecular Formula C8H8O3S
Quantity 25 g
Synonym 2-Oxo-2-(thiophen-2-yl)acetic Acid Ethyl Ester, Ethyl 2-oxo-2-(2-Thienyl)acetate, Ethyl 2-oxo-2-(2-Thienyl)ethanoate, Ethyl 2-oxo-2-(2'-thienyl)acetate, Ethyl 2-oxo-2-(Thiophen-2-yl)acetate, Ethyl 2-oxo-2-Thienylacetate, Ethyl 2-Thienylglyoxylate, Ethyl 2-Thiopheneglyoxylate, Ethyl oxo(2-Thienyl)acetate, Ethyl alpha-oxo-2-Thiopheneacetate
InChI Formula InChI=1S/C8H8O3S/c1-2-11-8(10)7(9)6-4-3-5-12-6/h3-5H,2H2,1H3
SMILES CCOC(=O)C(=O)c1cccs1
Molecular Weight (g/mol) 184.21
Formula Weight 184.0194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.