missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-2-glyoxylate, 97%

Product Code. p-7027727
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11490598

Brand: Thermo Scientific Alfa Aesar B24712.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4075-58-5 | C8H8O3S | 184.209 g/mol

Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4075-58-5
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.209
MDL Number MFCD00015538
InChI Key GHOVLEQTRNXASK-UHFFFAOYSA-N
Synonym ethyl thiophene-2-glyoxylate, ethyl 2-oxo-2-thiophen-2-yl acetate, ethyl 2-oxo-2-2-thienyl acetate, ethyl alpha-oxothiophen-2-acetate, ethyl 2-thienylglyoxylate, thiophen-2-coco-o-ethyl, 2-thiophene glyoxylic acid ethyl ester, oxo-thiophen-2-yl-acetic acid ethyl ester, 2-thiophene glyoxalicacid ethyl ester, ethyl oxo 2-thienyl acetate
PubChem CID 77693
IUPAC Name ethyl 2-oxo-2-thiophen-2-ylacetate
SMILES CCOC(=O)C(=O)C1=CC=CS1

Specifications

Density 1.25
Boiling Point 102°C to 110°C (0.8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5505
Quantity 10 g
Beilstein 5651
Solubility Information Difficult to mix.
Formula Weight 184.21
Percent Purity 97%
Chemical Name or Material Ethyl thiophene-2-glyoxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.