missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-2-glyoxylate, 97%

Product Code. 11490598
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490598 10 g 10g
11400608 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490598 Supplier Thermo Scientific Alfa Aesar Supplier No. B24712.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4075-58-5 | C8H8O3S | 184.209 g/mol

Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4075-58-5
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.209
MDL Number MFCD00015538
InChI Key GHOVLEQTRNXASK-UHFFFAOYSA-N
Synonym ethyl thiophene-2-glyoxylate, ethyl 2-oxo-2-thiophen-2-yl acetate, ethyl 2-oxo-2-2-thienyl acetate, ethyl alpha-oxothiophen-2-acetate, ethyl 2-thienylglyoxylate, thiophen-2-coco-o-ethyl, 2-thiophene glyoxylic acid ethyl ester, oxo-thiophen-2-yl-acetic acid ethyl ester, 2-thiophene glyoxalicacid ethyl ester, ethyl oxo 2-thienyl acetate
PubChem CID 77693
IUPAC Name ethyl 2-oxo-2-thiophen-2-ylacetate
SMILES CCOC(=O)C(=O)C1=CC=CS1

Specifications

Density 1.25
Boiling Point 102°C to 110°C (0.8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5505
Quantity 10 g
Beilstein 5651
Solubility Information Difficult to mix.
Formula Weight 184.21
Percent Purity 97%
Chemical Name or Material Ethyl thiophene-2-glyoxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.