missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-2-carboxylate, 98+%

Product Code. 11488920
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488920 10 g 10g
11498920 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488920 Supplier Thermo Scientific Alfa Aesar Supplier No. A12302.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2810-04-0 | C7H8O2S | 156.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2810-04-0
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.20
MDL Number MFCD00005436
InChI Key JZGZKRJVTIRPOK-UHFFFAOYSA-N
PubChem CID 76052
IUPAC Name ethyl thiophene-2-carboxylate
SMILES CCOC(=O)C1=CC=CS1

Specifications

Density 1.161
Boiling Point 217°C to 218°C
Flash Point 88°C (190°F)
Refractive Index 1.526
Quantity 10 g
Beilstein 112678
Synonym ethyl 2-thiophenecarboxylate, ethyl 2-thenoate, ethyl-2-thiophenecarboxylate, thiophene-2-carboxylic acid ethyl ester, 2-thiophenecarboxylic acid ethyl ester, thiophene, 2-ethyl carboxylate, thiophenecarboxylic acid, ethyl ester, ethylthiophen-2-carboxylat, ethyl2-thiophenecarboxylate, acmc-209h0w
Formula Weight 156.2
Percent Purity ≥98%
Chemical Name or Material Ethyl thiophene-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.