missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiooxamate, 95%

Product Code. 15494087
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494087 5 g 5g
15484087 1 g 1g
15404097 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15494087 Supplier Thermo Scientific Alfa Aesar Supplier No. H31447.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16982-21-1 | C4H7NO2S | 133.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16982-21-1
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00074903
InChI Key YMBMCMOZIGSBOA-UHFFFAOYSA-N
Synonym ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
PubChem CID 2733398
IUPAC Name ethyl 2-amino-2-sulfanylideneacetate
SMILES CCOC(=O)C(=S)N

Specifications

Melting Point 61°C to 65°C
Quantity 5 g
Formula Weight 133.17
Percent Purity 95%
Chemical Name or Material Ethyl thiooxamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.