missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tetrahydropyran-4-ylacetate, 97%, Thermo Scientific™

Product Code. 10012504
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10012504 5 g Amber Glass Bottle 5g
10421934 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10012504 Supplier Thermo Scientific Maybridge Supplier No. CC67423DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 103260-44-2
Molecular Formula C9H16O3
Molecular Weight (g/mol) 172.224
InChI Key JLMMMEDWRUVCEW-UHFFFAOYSA-N
Synonym ethyl tetrahydropyran-4-ylacetate, ethyl 2-tetrahydro-2h-pyran-4-yl acetate, ethyl 2-oxan-4-yl acetate, ethyl tetrahydropyran-4-yl-acetate, ethyl 2-4-tetrahydropyranyl acetate, ethyl tetrahydropyranyl-4-acetate, 2h-pyran-4-acetic acid, tetrahydro-, ethyl ester, ethyl tetrahydropyran-4-acetate, tetrahydro-pyran-4-yl-acetic acid ethyl ester, ethyl tetrahydro-2h-pyran-4-ylacetate
PubChem CID 2773412
IUPAC Name ethyl 2-(oxan-4-yl)acetate
SMILES CCOC(=O)CC1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl tetrahydropyran-4-ylacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.