missing translation for 'onlineSavingsMsg'
Learn More

Ethyl succinyl chloride, 95%

Product Code. 10772611 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10772611 100 g Glass bottle 100g
10489670 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772611 Supplier Thermo Scientific Acros Supplier No. 184091000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14794-31-1 | C6H9ClO3 | 164.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14794-31-1
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000751
InChI Key IXZFDJXHLQQSGQ-UHFFFAOYSA-N
Synonym ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride
PubChem CID 84648
IUPAC Name ethyl 4-chloro-4-oxobutanoate
SMILES CCOC(=O)CCC(=O)Cl

Specifications

Color Colorless to Yellow
Density 1.1575g/mL
Boiling Point 88.0°C to 90.0°C (11.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCH2CH2CO2C2H5
Refractive Index 1.4350 to 1.4390
Quantity 100 g
Beilstein 02, 613
Specific Gravity 1.1575
Formula Weight 164.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl succinyl chloride, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl succinyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.