missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Succinoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Succinoyl Chloride |
| CAS | 14794-31-1 |
| Molecular Formula | C6H9ClO3 |
| Quantity | 50 g |
| Synonym | 3-(Ethoxycarbonyl)propanoyl Chloride, 3-(Ethoxycarbonyl)propionyl Chloride, 3-Carbethoxypropionyl Chloride, 3-Chlorocarbonylpropionic Acid Ethyl Ester, 4-Chloro-4-oxobutanoic Acid Ethyl Ester, 4-Ethoxy-4-oxobutanoic Acid Chloride, Ethyl 3-(chlorocarbonyl)propanoate, Ethyl 3-(chloroformyl)propionate, Ethyl 3-Chlorocarbonylpropionate, Ethyl 4-chloro-4-oxobutanoate, Ethyl 4-Chloro-4-oxobutyrate, Ethyl Succinoyl Chloride, Ethyl Succinoyl Monochloride, Ethyl Succinyl Chloride, Monoethyl Succinoyl Chloride, Succinic Acid Chloride Monoethyl Ester, Succinic Acid Ethyl Ester Chloride, Succinic Acid Monoethyl Chloride, beta-Carbethoxypropionyl Chloride, beta-Ethoxycarbonylpropionyl Chloride |
| InChI Formula | InChI=1S/C6H9ClO3/c1-2-10-6(9)4-3-5(7)8/h2-4H2,1H3 |
| SMILES | CCOC(=O)CCC(=O)Cl |
| IUPAC Name | ethyl 4-chloro-4-oxobutanoate |
| Molecular Weight (g/mol) | 164.59 |
| Formula Weight | 164.024 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?