missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Succinoyl Chloride, TRC

Product Code. 30374174
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30374174 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30374174

Brand: Toronto Research Chemicals TRCE818230100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Succinoyl Chloride
CAS 14794-31-1
Molecular Formula C6H9ClO3
Quantity 100 g
Synonym 3-(Ethoxycarbonyl)propanoyl Chloride, 3-(Ethoxycarbonyl)propionyl Chloride, 3-Carbethoxypropionyl Chloride, 3-Chlorocarbonylpropionic Acid Ethyl Ester, 4-Chloro-4-oxobutanoic Acid Ethyl Ester, 4-Ethoxy-4-oxobutanoic Acid Chloride, Ethyl 3-(chlorocarbonyl)propanoate, Ethyl 3-(chloroformyl)propionate, Ethyl 3-Chlorocarbonylpropionate, Ethyl 4-chloro-4-oxobutanoate, Ethyl 4-Chloro-4-oxobutyrate, Ethyl Succinoyl Chloride, Ethyl Succinoyl Monochloride, Ethyl Succinyl Chloride, Monoethyl Succinoyl Chloride, Succinic Acid Chloride Monoethyl Ester, Succinic Acid Ethyl Ester Chloride, Succinic Acid Monoethyl Chloride, beta-Carbethoxypropionyl Chloride, beta-Ethoxycarbonylpropionyl Chloride
InChI Formula InChI=1S/C6H9ClO3/c1-2-10-6(9)4-3-5(7)8/h2-4H2,1H3
SMILES CCOC(=O)CCC(=O)Cl
IUPAC Name ethyl 4-chloro-4-oxobutanoate
Molecular Weight (g/mol) 164.59
Formula Weight 164.024
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.