missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Stearate, TRC

Product Code. 17994705
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17994705 1 g 1g
17952035 100 mg 100mg
17993195 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17994705 Supplier Toronto Research Chemicals Supplier No. TRCE9259501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-61-5
Molecular Formula C20 H40 O2
Molecular Weight (g/mol) 312.53
Synonym Ethyl Stearate, Octadecanoic acid ethyl ester, Octadecanoic acid, ethyl ester (9CI, ACI), Stearic acid, ethyl ester (6CI, 8CI), Ethyl n-octadecanoate, Ethyl octadecanoate, Ethyl stearate, KAK-ES, NSC 8919
IUPAC Name ethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H40O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h3-19H2,1-2H3
Formula Weight 312.3028
Chemical Name or Material Octadecanoic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.