missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Stearate-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Stearate-d5 |
| CAS | 1216808-22-8 |
| Molecular Formula | C20 D5 H35 O2 |
| Quantity | 50 mg |
| Synonym | 1,1,2,2,2-Pentadeuterioethyl octadecanoate, Octadecanoic Acid Ethyl Ester-d5, Stearic Acid Ethyl Ester-d5, Ethyl n-Octadecanoate-d5, Ethyl Octadecanoate-d5, KAK-ES-d5, NSC 8919-d5 |
| InChI Formula | InChI=1S/C20H40O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h3-19H2,1-2H3/i2D3,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])OC(=O)CCCCCCCCCCCCCCCCC |
| IUPAC Name | 1,1,2,2,2-pentadeuterioethyl octadecanoate |
| Molecular Weight (g/mol) | 317.5612 |
| Formula Weight | 317.3342 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?