missing translation for 'onlineSavingsMsg'
Learn More

Ethyl sorbate, 96%

Product Code. 11420283
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420283 25 g 25g
11430283 100 g 100g
11440283 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420283 Supplier Thermo Scientific Alfa Aesar Supplier No. A11431.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2396-84-1 | C8H12O2 | 140.18 g/mol

The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The addition of chlorine and bromine to ethyl sorbate (la) gave 1, 2-and 1, a-dihalo products derived from attack of the halogen. Chlorination of la under ionic conditions proceeds through a tightly bridged chloronium ion intermediate.

Solubility
Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2396-84-1
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00009296
InChI Key OZZYKXXGCOLLLO-TWTPFVCWSA-N
Synonym ethyl sorbate, ethyl 2,4-hexadienoate, ethyl hexa-2,4-dienoate, sorbic acid, ethyl ester, 2,4-hexadienoic acid, ethyl ester, ethyl 2e,4e-hexa-2,4-dienoate, ethyl e,e-2,4-hexadienoate, unii-hsr16usg4d, 2,4-hexadienoic acid, ethyl ester, 2e,4e, fema no. 2459
PubChem CID 1550470
IUPAC Name ethyl (2E,4E)-hexa-2,4-dienoate
SMILES CCOC(=O)\C=C\C=C\C

Specifications

Density 0.948
Boiling Point 195°C to 196°C
Flash Point 69°C (156°F)
Refractive Index 1.495
Quantity 25 g
Beilstein 1362941
Solubility Information Soluble in alcohol.
Formula Weight 140.18
Percent Purity 98%
Chemical Name or Material Ethyl sorbate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.