missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%

Product Code. 11349126
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349126 25 g 25g
11339126 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349126 Supplier Thermo Scientific Alfa Aesar Supplier No. L20259.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 312745-91-8 | C7H11NO3 | 157.17 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312745-91-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.17
MDL Number MFCD00672878
InChI Key LOQFROBMBSKWQY-UHFFFAOYNA-N
Synonym ethyl s-4-cyano-3-hydroxybutyrate, s-ethyl 4-cyano-3-hydroxybutanoate, ethyl 3s-4-cyano-3-hydroxybutanoate, s-ethyl-4-cyano-3-hydroxybutyrate, ethyl s-+-4-cyano-3-hydroxybutyrate, butanoic acid, 4-cyano-3-hydroxy-, ethyl ester, 3s, 4-cyano-3-hydroxy-butyric acid ethyl ester, pubchem21112
PubChem CID 7020868
IUPAC Name ethyl (3S)-4-cyano-3-hydroxybutanoate
SMILES CCOC(=O)CC(O)CC#N

Specifications

Density 1.114
Boiling Point 270°C
Flash Point >110°C (230°F)
Refractive Index 1.448
Quantity 25 g
Solubility Information Not miscible or difficult to mix with water.
Optical Rotation +5° (Neat)
Formula Weight 157.17
Percent Purity 97%
Assay ee ≥98%
Chemical Name or Material Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.