missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate, 97%, ACROS Organics™

Brand:  Acros Organics 361290010

Code : 29

Additional Details : CAS Number : 86728-85-0 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10292902

  • £23.00 / 1g


EU Stock 5
Add to basket


Specifications

Chemical Identifiers

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate
≥96.0%
C6H11ClO3
MFCD00211241
86728-85-0
97%
166.61

Specifications

Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate
86728-85-0
Authentic
1.4515 to 1.4535 (20°C, 589nm)
− 14.50 (20.00°C c=neat)
Liquid
97%
166.61
1g
1.1900g/mL
Glass bottle
-13° to -16° (20°C, 589nm) (neat)
Clear colorless to yellow
≥96.0%
C6H11ClO3
MFCD00211241
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl (S)-(-)-4-chloro-3-hydroxybutyrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS