missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(+)-3-hydroxybutyrate, Thermo Scientific Chemicals

Product Code. 15415449
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415449 1 g 1g
15425449 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415449 Supplier Thermo Scientific Alfa Aesar Supplier No. J63184.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For synthesis of optically active products

Ethyl (S)-(+)-3-hydroxybutyrate is used for synthesis of optically active products, used as medical and organic intermediates, an important amino protective agents, pharmaceuticals and synthetic organic makes intermediates, organic synthesis used to introduce t-Boc protect gene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl (S)-(+)-3-hydroxybutyrate is used for synthesis of optically active products, used as medical and organic intermediates, an important amino protective agents, pharmaceuticals and synthetic organic makes intermediates, organic synthesis used to introduce t-Boc protect gene.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56816-01-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00066206
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
Synonym s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
PubChem CID 6950306
IUPAC Name ethyl (3S)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Colorless to Yellow
Quantity 1 g
Solubility Information Soluble in chloroform
Formula Weight 132.16
Physical Form Liquid
Chemical Name or Material Ethyl (S)-(+)-3-hydroxybutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.