missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-3-hydroxybutyrate, 99%, Thermo Scientific™

Product Code. 10011701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10011701 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10011701 Supplier Thermo Scientific Acros Supplier No. 335260010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 56816-01-4
Color Colorless
Density 1.0100g/mL
Boiling Point 180°C to 182°C
Flash Point 64°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12O3
Linear Formula CH3CH(OH)CH2COOCH2CH3
Refractive Index 1.4200 to 1.422
MDL Number MFCD00066206
Quantity 1 g
Specific Gravity 1.01
Specific Rotation Condition + 43.00 (20.00°C c=1,chloroform)
Specific Rotation + 43
Synonym s-ethyl 3-hydroxybutanoate, ethyl s-3-hydroxybutyrate, ethyl 3s-3-hydroxybutanoate, s-+-3-hydroxybutyric acid ethyl ester, ethyl s-+-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid ethyl ester, ethyl s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, ethyl ester, 3s, ethyl s---3-hydroxybutyrate
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
SMILES CCOC(=O)CC(C)O
IUPAC Name ethyl (3S)-3-hydroxybutanoate
Molecular Weight (g/mol) 132.16
PubChem CID 6950306
Formula Weight 132.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl (S)-3-hydroxybutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.