missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-(-)-2-methoxypropionate, 98%, Thermo Scientific Chemicals

Product Code. 10205581 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205581 1 mL Glass bottle 1mL
10396841 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205581 Supplier Thermo Scientific Acros Supplier No. 331730010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41918-08-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD01632590
InChI Key WHRLOJCOIKOQGL-YFKPBYRVSA-N
Synonym ethyl 2s-2-methoxypropanoate, s-ethyl 2-methoxypropanoate, ethyl s-2-methoxypropanoate, ethyl s---2-methoxypropionate, 2-methoxypropionic acid ethyl ester, s, ?-2-methoxypropionic acid ethyl ester, propanoic acid,2-methoxy-, ethyl ester, 2s
PubChem CID 5702567
IUPAC Name ethyl (2S)-2-methoxypropanoate
SMILES CCOC(=O)C(C)OC

Specifications

Color Colorless to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OCH3)COOCH2CH3
Refractive Index 1.3970 to 1.3990
Quantity 1 mL
Beilstein 03, III, 446
Specific Rotation Condition − 89.00 (20.00°C c=neat)
Specific Rotation − 89.00
Formula Weight 132.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl (S)-(-)-2-methoxypropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.