missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (S)-1-Phenyl-1,2,3,4-tetrahydro-2-isoquinolinecarboxylate, TRC

Product Code. 17934105
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17934105 100 mg 100mg
17912495 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17934105 Supplier Toronto Research Chemicals Supplier No. TRCE679700100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180468-42-2
Molecular Formula C18 H19 N O2
Molecular Weight (g/mol) 281.35
Synonym (1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic acid ethyl ester, 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, ethyl ester, (S)-, (S)-1-Phenyl-3,4-dihydro-1H-isoquinolinecarboxylic acid ethyl ester, Ethyl (1S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate, Ethyl (1S)-3,4-dihydro-1-phenyl-2(1H)-isoquinolinecarboxylate, Ethyl (S)-1-phenyl-1,2,3,4-tetrahydro-2-isoquinolinecarboxylate, Ethyl (S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
IUPAC Name ethyl (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate
SMILES CCOC(=O)N1CCc2ccccc2[C@@H]1c3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H19NO2/c1-2-21-18(20)19-13-12-14-8-6-7-11-16(14)17(19)15-9-4-3-5-10-15/h3-11,17H,2,12-13H2,1H3/t17-/m0/s1
Formula Weight 281.1416
Chemical Name or Material Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.