missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Red, pure, ACROS Organics™

Product Code. 10286960 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10286960 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10286960 Supplier Thermo Scientific Acros Supplier No. 190090010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 76058-33-8
Molecular Formula C17H18N3O2
MDL Number MFCD00002427
Beilstein 16, I, 316
Synonym 2-[4-(Diethylamino)phenylazo]benzoic acid
InChI Key HBRCDTRQDHMTDA-UHFFFAOYSA-M
Infrared Spectrum Authentic
SMILES CCN(CC)C1=CC=C(C=C1)N=NC1=CC=CC=C1C([O-])=O
IUPAC Name 2-{2-[4-(diethylamino)phenyl]diazen-1-yl}benzoate
Molecular Weight (g/mol) 296.35
PubChem CID 54832
Formula Weight 297.35
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 4.4 (red) to pH 6.2 (yellow)
Color Red
Quantity 1 g
Physical Form Powder
Chemical Name or Material Ethyl Red
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.