missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-(-)-3-hydroxybutyrate, Thermo Scientific™

Product Code. 15455649
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455649 1 g 1g
15465649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455649 Supplier Thermo Scientific Alfa Aesar Supplier No. J63400.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For synthesis of optically active products

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24915-95-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00075386
InChI Key OMSUIQOIVADKIM-RXMQYKEDSA-N
Synonym ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester
PubChem CID 440030
ChEBI CHEBI:28707
IUPAC Name ethyl (3R)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Colorless
Quantity 1 g
Formula Weight 132.16
Physical Form Liquid
Chemical Name or Material Ethyl (R)-(-)-3-hydroxybutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.