missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-3-hydroxybutyrate, 98%

Product Code. 10397701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10397701 1 g Glass bottle 1g
10441392 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10397701 Supplier Thermo Scientific Acros Supplier No. 345760010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24915-95-5 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24915-95-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00075386
InChI Key OMSUIQOIVADKIM-RXMQYKEDSA-N
Synonym ethyl r-3-hydroxybutyrate, ethyl 3r-3-hydroxybutanoate, ethyl r-3-hydroxybutanoate, ethyl r---3-hydroxybutyrate, r-ethyl 3-hydroxybutanoate, r---3-hydroxybutyric acid ethyl ester, r-3-hydroxybutyric acid ethyl ester, butanoic acid, 3-hydroxy-, ethyl ester, 3r, r---3-hydroxy-n-butyric acid ethyl ester
PubChem CID 440030
ChEBI CHEBI:28707
IUPAC Name ethyl (3R)-3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Color Colorless
Density 1.0170g/mL
Boiling Point 75°C to 76°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH2COOCH2CH3
Refractive Index 1.4195 to 1.4215
Quantity 1 g
Specific Gravity 1.017
Specific Rotation Condition −45.5° (20°C 589nm,c=1,CHCl3)
Specific Rotation -45.5
Solubility Information Solubility in water: soluble in water
Formula Weight 132.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl (R)-3-hydroxybutyrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.