missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyruvate, 98%

Product Code. 10764041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10764041 100 g Glass bottle 100g
10408650 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10764041 Supplier Thermo Scientific Acros Supplier No. 118941000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-35-6 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-35-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00009123
InChI Key XXRCUYVCPSWGCC-UHFFFAOYSA-N
Synonym ethyl pyruvate, ethyl 2-oxopropionate, pyruvic acid, ethyl ester, propanoic acid, 2-oxo-, ethyl ester, pyruvic acid ethyl ester, ethyl pyroracemate, ethyl acetylformate, 2-oxo-propionic acid ethyl ester, ethyl pyruvate natural, ethyl alpha-ketopropionate
PubChem CID 12041
IUPAC Name ethyl 2-oxopropanoate
SMILES CCOC(=O)C(=O)C

Specifications

Melting Point -58.0°C
Color Yellow
Density 1.0453g/mL
Boiling Point 148.0°C to 150.0°C
Flash Point 45°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3COCO2C2H5
Refractive Index 1.4040 to 1.4060
Quantity 100 g
Beilstein 03, 616
Merck Index 15, 8135
Specific Gravity 1.0453
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethanol,ether
Viscosity 1.18 mPa.s
Formula Weight 116.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl pyruvate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl pyruvate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl pyruvate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.