missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyrrolidinoacetate, 98%, Thermo Scientific™

Catalog Number 10478671
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
22041-19-6
C8H15NO2
157.213
MFCD00012085
IIIBKVMTKMLDFJ-UHFFFAOYSA-N
ethyl 2-pyrrolidin-1-yl acetate, ethyl 1-pyrrolidineacetate, ethylpyrrolidinoacetate, 1-pyrrolidineacetic acid, ethyl ester, ethyl pyrrolidinoacetate, ethyl pyrrolidin-1-ylacetate, ethyl n-pyrrolidinylacetate, ethyl 1-pyrrolidinylacetate, pyrrolidin-1-yl-acetic acid ethyl ester, ethyl-1-pyrrolidineacetate
210996
ethyl 2-pyrrolidin-1-ylacetate
CCOC(=O)CN1CCCC1
This item is not returnable. View return policy
22041-19-6
C8H15NO2
157.213
MFCD00012085
IIIBKVMTKMLDFJ-UHFFFAOYSA-N
ethyl 2-pyrrolidin-1-yl acetate, ethyl 1-pyrrolidineacetate, ethylpyrrolidinoacetate, 1-pyrrolidineacetic acid, ethyl ester, ethyl pyrrolidinoacetate, ethyl pyrrolidin-1-ylacetate, ethyl n-pyrrolidinylacetate, ethyl 1-pyrrolidinylacetate, pyrrolidin-1-yl-acetic acid ethyl ester, ethyl-1-pyrrolidineacetate
210996
ethyl 2-pyrrolidin-1-ylacetate
CCOC(=O)CN1CCCC1
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Quantity 5 g
Beilstein 20, V,1, 335
Formula Weight 157.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl pyrrolidinoacetate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'