missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Propionylacetate, TRC

Product Code. 30387722
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387722 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387722 Supplier Toronto Research Chemicals Supplier No. TRCE92539510G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Propionylacetate
CAS 4949-44-4
Molecular Formula C7H12O3
Quantity 10 g
Synonym 3-Oxopentanoic Acid Ethyl Ester, 3-Oxovaleric Acid Ethyl Ester, 3-Oxopentanoic Acid Ethyl Ester, Ethyl 3-Ketopentanoate, Ethyl 3-Oxopentanoate, Ethyl 3-Oxovalerate, Ethyl Propanoylacetate, Ethyl Propioacetate, Ethyl .beta.-oxovalerate, NSC 7251, NSC 78662, Propionylacetic Acid Ethyl Ester
InChI Formula InChI=1S/C7H12O3/c1-3-6(8)5-7(9)10-4-2/h3-5H2,1-2H3
SMILES CCOC(=O)CC(=O)CC
IUPAC Name ethyl 3-oxopentanoate
Molecular Weight (g/mol) 144.17
Formula Weight 144.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.