missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl propionylacetate, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 4949-44-4 |
---|---|
Molecular Formula | C7H12O3 |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD00009317 |
InChI Key | UDRCONFHWYGWFI-UHFFFAOYSA-N |
Synonym | ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate |
PubChem CID | 78656 |
IUPAC Name | ethyl 3-oxopentanoate |
SMILES | CCC(=O)CC(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10229213
|
Thermo Scientific Acros
232910010 |
1 mL | Glass bottle |
N/A
|
N/A | ||||
10751701
|
Thermo Scientific Acros
232910050 |
5 mL | Glass bottle |
N/A
|
N/A | ||||
10296760
|
Thermo Scientific Acros
232910250 |
25 mL | Glass bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4949-44-4 | |
144.17 | |
UDRCONFHWYGWFI-UHFFFAOYSA-N | |
78656 | |
CCC(=O)CC(=O)OCC |
C7H12O3 | |
MFCD00009317 | |
ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate | |
ethyl 3-oxopentanoate |
Specifications
4949-44-4 | |
1.0100g/mL | |
77°C | |
98% min. (GC) | |
C2H5COCH2CO2C2H5 | |
MFCD00009317 | |
ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate | |
CCC(=O)CC(=O)OCC | |
144.17 | |
144.17 | |
Liquid |
Colorless to Yellow | |
83.0°C to 84.0°C (12.0 mmHg) | |
Authentic | |
C7H12O3 | |
1.4220 to 1.4240 | |
1.01 | |
UDRCONFHWYGWFI-UHFFFAOYSA-N | |
ethyl 3-oxopentanoate | |
78656 | |
98+% | |
Ethyl propionylacetate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 225-593-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethyl propionylacetate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only