missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propionylacetate, 95%

Product Code. 11482827
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482827 100 g 100g
11462827 5 g 5g
11472827 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482827 Supplier Thermo Scientific Alfa Aesar Supplier No. A15464.22
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4949-44-4 | C7H12O3 | 144.17 g/mol

Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.

Solubility
Miscible with glacial acetic acid.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4949-44-4
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009317
InChI Key UDRCONFHWYGWFI-UHFFFAOYSA-N
Synonym ethyl propionylacetate, ethyl 3-oxovalerate, ethyl 3-oxo-n-valerate, pentanoic acid, 3-oxo-, ethyl ester, 3-oxopentanoic acid ethyl ester, 3-oxo-pentanoic acid ethyl ester, ethyl 3-ketopentanoate, 3-oxovaleric acid ethyl ester, propionylacetic acid ethyl ester, ethylpropanylacetate
PubChem CID 78656
IUPAC Name ethyl 3-oxopentanoate
SMILES CCC(=O)CC(=O)OCC

Specifications

Density 1.012
Boiling Point 190°C to 192°C
Flash Point 77°C (170°F)
Refractive Index 1.422
Quantity 100 g
Beilstein 970270
Solubility Information Miscible with glacial acetic acid.
Formula Weight 144.17
Percent Purity 95%
Chemical Name or Material Ethyl propionylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.