missing translation for 'onlineSavingsMsg'
Learn More

Ethyl propenyl ether, 98%, mixture of cis- and trans-isomers

Product Code. 10177183 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177183 10 g Glass bottle 10g
10493351 25 g Glass bottle 25g
10475872 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10177183 Supplier Thermo Scientific Acros Supplier No. 410370100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-55-2 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-55-2
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00015184
InChI Key XDHOEHJVXXTEDV-HYXAFXHYSA-N
Synonym 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
PubChem CID 5365091
IUPAC Name (E)-1-ethoxyprop-1-ene
SMILES CCO\C=C/C

Specifications

Color Colorless to Yellow
Density 0.7780g/mL
Boiling Point 67.0°C to 76.0°C
Flash Point −18°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CHCHOCH2CH3
Refractive Index 1.3940 to 1.4000
Quantity 10 g
Fieser 01,385
Specific Gravity 0.778
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl propenyl ether, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl propenyl ether, mixture of cis- and trans-isomers
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.