missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Propargylate, TRC

Product Code. 30372885
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372885 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372885 Supplier Toronto Research Chemicals Supplier No. TRCE81806025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Ethyl Propargylate
CAS 623-47-2
Molecular Formula C5H6O2
Quantity 25 g
Synonym (Ethoxycarbonyl)acetylene, Carboethoxyacetylene, Ethyl 2-Propynoate, Ethyl Acetylenecarboxylate, Ethyl Acetylenemonocarboxylate, Ethyl Propiolate, Ethyl Propynoate, NSC 60551, Propynoic Acid Ethyl Ester
InChI Formula InChI=1S/C5H6O2/c1-3-5(6)7-4-2/h1H,4H2,2H3
SMILES CCOC(=O)C#C
IUPAC Name ethyl prop-2-ynoate
Molecular Weight (g/mol) 98.1
Formula Weight 98.0368
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.