missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Propargylate, TRC

Product Code. 30382609
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382609 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382609 Supplier Toronto Research Chemicals Supplier No. TRCE8180605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Propargylate
CAS 623-47-2
Molecular Formula C5H6O2
Quantity 5 g
Synonym (Ethoxycarbonyl)acetylene, Carboethoxyacetylene, Ethyl 2-Propynoate, Ethyl Acetylenecarboxylate, Ethyl Acetylenemonocarboxylate, Ethyl Propiolate, Ethyl Propynoate, NSC 60551, Propynoic Acid Ethyl Ester
InChI Formula InChI=1S/C5H6O2/c1-3-5(6)7-4-2/h1H,4H2,2H3
SMILES CCOC(=O)C#C
IUPAC Name ethyl prop-2-ynoate
Molecular Weight (g/mol) 98.1
Formula Weight 98.0368
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.