missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Propargylate, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Ethyl Propargylate |
| CAS | 623-47-2 |
| Molecular Formula | C5H6O2 |
| Quantity | 10 g |
| Synonym | (Ethoxycarbonyl)acetylene, Carboethoxyacetylene, Ethyl 2-Propynoate, Ethyl Acetylenecarboxylate, Ethyl Acetylenemonocarboxylate, Ethyl Propiolate, Ethyl Propynoate, NSC 60551, Propynoic Acid Ethyl Ester |
| InChI Formula | InChI=1S/C5H6O2/c1-3-5(6)7-4-2/h1H,4H2,2H3 |
| SMILES | CCOC(=O)C#C |
| IUPAC Name | ethyl prop-2-ynoate |
| Molecular Weight (g/mol) | 98.1 |
| Formula Weight | 98.0368 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction