missing translation for 'onlineSavingsMsg'
Learn More

Ethyl picolinoylacetate, 95%

Product Code. 10657474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657474 10 g Glass bottle 10g
10259420 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657474 Supplier Thermo Scientific Acros Supplier No. 425470100
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 26510-52-1 | C10H11NO3 | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26510-52-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094022
InChI Key FQHXWZMJALFSJJ-UHFFFAOYSA-N
Synonym ethyl 3-oxo-3-pyridin-2-yl propanoate, ethyl picolinoylacetate, ethyl 3-oxo-3-2-pyridyl propionate, ethyl 3-oxo-3-2-pyridinyl propanoate, ethyl 3-oxo-3-2-pyridyl propanoate, pubchem11088, ethyl-2-pyridoyl acetate, d05hbz, ethyl 3-pyridinyl-3-oxopropionate, 2-3-ethoxy-3-oxopropanoyl pyridine
PubChem CID 2736461
IUPAC Name ethyl 3-oxo-3-pyridin-2-ylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=N1

Specifications

Color Brown to Straw-Yellow
Density 1.1640g/mL
Boiling Point 100°C to 104°C (0.3 mmHg)
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5160 to 1.519
Quantity 10 g
Beilstein 22,305
Specific Gravity 1.164
Solubility Information Solubility in water: negligible
Formula Weight 193.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl picolinoylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.