missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Phenylcyanoacetate, TRC

Product Code. 30385055
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30385055 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30385055

Brand: Toronto Research Chemicals TRCE92600510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Phenylcyanoacetate
CAS 4553-07-5
Molecular Formula C11H11NO2
Quantity 10 g
Synonym Cyanophenyl-acetic Acid Ethyl Ester, Ethyl (±)-2-cyano-2-phenylacetate, Ethyl 2-cyano-2-phenylacetate, Ethyl 2-phenyl-2-cyanoacetate, Ethyl Cyanophenylacetate, Ethyl alpha-cyano-alpha-phenylacetate, Ethyl alpha-cyanobenzeneacetate, Ethyl alpha-cyanophenylacetate, NSC 362, Phenylcyanoacetic Acid Ethyl Ester
InChI Formula InChI=1S/C11H11NO2/c1-2-14-11(13)10(8-12)9-6-4-3-5-7-9/h3-7,10H,2H2,1H3
SMILES CCOC(=O)C(C#N)c1ccccc1
Molecular Weight (g/mol) 189.21
Formula Weight 189.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.