missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Phenylacetate, TRC
Specifications
Specifications
| Chemical Name or Material | Phenylacetic Acid Ethylester |
| CAS | 101-97-3 |
| Molecular Formula | C10 H12 O2 |
| Quantity | 25 g |
| Synonym | Ethyl 2-phenyl Acetate, Ethyl 2-phenylacetate, Benzeneacetic acid, ethyl ester, Acetic acid, phenyl-, ethyl ester (6CI,8CI), 2-Phenylacetic acid ethyl ester, Ethyl 2-phenylacetate, Ethyl 2-phenylethanoate, Ethyl benzeneacetate, Ethyl phenacetate, Ethyl phenylacetate, Ethyl α-phenylacetate, Ethyl α-toluate, NSC 406259, NSC 8894, Phenylacetic acid ethyl ester, α-Toluic acid ethyl ester |
| InChI Formula | InChI=1S/C10H12O2/c1-2-12-10(11)8-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3 |
| SMILES | CCOC(=O)Cc1ccccc1 |
| IUPAC Name | ethyl 2-phenylacetate |
| Molecular Weight (g/mol) | 164.2 |
| Formula Weight | 164.0837 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?