missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylacetate, 99%

Product Code. 10205440 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10205440

Brand: Thermo Scientific Acros 148432500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 101-97-3 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-97-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
InChI Key DULCUDSUACXJJC-UHFFFAOYSA-N
IUPAC Name ethyl 2-phenylacetate
SMILES CCOC(=O)CC1=CC=CC=C1

Specifications

Melting Point -29.0°C
Density 1.0300 g/mL
Boiling Point 226.0°C
Flash Point 102°C
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5CH2CO2C2H5
Refractive Index 1.4970 to 1.4990
Quantity 250 g
Beilstein 09, 434
Merck Index 15, 3893
Specific Gravity 1.03
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: very soluble in ethanol and diethyl ether, soluble in chloroform
Formula Weight 164.2
Chemical Name or Material Ethyl phenylacetate
Show More Show Less
Product Identifier
  • Ethyl phenylacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.