missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Palmitoleate-d5, TRC

Product Code. 30380758
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380758 10 mg
30371672 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30380758 Supplier Toronto Research Chemicals Supplier No. TRCE92585710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C18H29D5O2
Molecular Weight (g/mol) 287.49
Synonym (9Z)-9-Hexadecenoic Acid Ethyl Ester-d5, (Z)-9-Hexadecenoic Acid Ethyl Ester-d5, Palmitoleic Acid Ethyl Ester-d5, Ethyl (Z)-9-Hexadecenoate-d5, Ethyl cis-9-Hexadecenoate-d5, Ethyl Palmitoleate-d5, cis-9-Hexadecenoic Acid Ethyl Ester-d5
SMILES O=C(CCCCCCC/C=C\CCCCCC)OC([2H])([2H])C([2H])([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h9-10H,3-8,11-17H2,1-2H3/b10-9-/i2D3,4D2
Chemical Name or Material Ethyl Palmitoleate-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.