missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Palmitate-d5, TRC

Product Code. 30382282
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382282 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30382282 Supplier Toronto Research Chemicals Supplier No. TRCE925482250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Palmitate-d5
CAS 1215397-47-9
Molecular Formula C18 D5 H31 O2
Quantity 250 mg
Synonym 1,1,2,2,2-Pentadeuterioethyl hexadecanoate, Hexadecanoic Acid Ethyl Ester-d5, Palmitic Acid Ethyl Ester-d5, Ethyl Hexadecanoate-d5, NSC 8918-d5
InChI Formula InChI=1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3/i2D3,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])OC(=O)CCCCCCCCCCCCCCC
IUPAC Name 1,1,2,2,2-pentadeuterioethyl hexadecanoate
Molecular Weight (g/mol) 289.508
Formula Weight 289.3029
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.