missing translation for 'onlineSavingsMsg'
Learn More

Ethyl palmitate, 1mg/ml in Acetonitrile, Lipomed

Product Code. 30459793
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459793 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459793 Supplier Lipomed Supplier No. LPMFAE14511LA
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Ethyl palmitate, 1mg/ml in Acetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylpalmitate
CAS 628-97-7
Analyte or Component Names Ethylpalmitate
Shipping Condition Room Temperature
Molecular Formula C18 H36 O2
Concentration 1 mg/mL
Synonym Ethyl Palmitate, Hexadecanoic acid ethyl ester
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CCCCCCCCCCCCCCCC(=O)OCC
IUPAC Name ethyl hexadecanoate
Solvent or Matrix Acetonitrile
Industry Type Pharmaceutical
Molecular Weight (g/mol) 284.48
Quantity 1 mL
Formula Weight 284.2715 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.