missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-toluenesulfonate, 98%

Product Code. 10756011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10756011 1 kg Glass bottle 1kg
10740595 50 g Glass bottle 50g
10478100 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10756011 Supplier Thermo Scientific Acros Supplier No. 147230010
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-40-0 | C9H12O3S | 200.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-40-0
Molecular Formula C9H12O3S
Molecular Weight (g/mol) 200.25
MDL Number MFCD00009100
InChI Key VRZVPALEJCLXPR-UHFFFAOYSA-N
Synonym ethyl p-toluenesulfonate, ethyl tosylate, ethyl p-tosylate, ethyl p-ts, benzenesulfonic acid, 4-methyl-, ethyl ester, ethyl p-methylbenzenesulfonate, ethyl-p-toluenesulfonate, p-toluenesulfonic acid, ethyl ester, ethyl pts, p-toluolsulfonsaeure aethyl ester
PubChem CID 6638
IUPAC Name ethyl 4-methylbenzenesulfonate
SMILES CCOS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Melting Point 32.0°C to 34.0°C
Color Colorless to Brown
Density 1.1740g/mL
Boiling Point 158.0°C to 162.0°C (10.0 mmHg)
Flash Point 157°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4SO3C2H5
Refractive Index 1.5100 to 1.5120
Quantity 1 kg
Beilstein 11, 99
Merck Index 15, 3907
Specific Gravity 1.174
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
Formula Weight 200.25
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Ethyl p-toluenesulfonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.