missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-aminobenzoate, 99.90%, MP Biomedicals™

Product Code. 11475411 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475411 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11475411 Supplier MP Biomedicals Supplier No. 0215796780
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 94-09-7
Assay Percent Range 0.999
Molecular Formula C9H11NO2
MDL Number MFCD00007892
Quantity 100 g
Synonym benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
InChI Key BLFLLBZGZJTVJG-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC=C(N)C=C1
IUPAC Name ethyl 4-aminobenzoate
Molecular Weight (g/mol) 165.19
PubChem CID 2337
ChEBI CHEBI:116735
Formula Weight 165.2
Physical Form Crystalline
Chemical Name or Material Ethyl p-aminobenzoate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.