missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Orange sodium salt, Thermo Scientific Chemicals

Product Code. 15405169
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405169 100 g 100g
15415169 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405169 Supplier Thermo Scientific Alfa Aesar Supplier No. J62863.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator: pH 3.0 (red) to pH 4.5 (yellow)

Indicator: pH 3.0 (red) to pH 4.5 (yellow)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62758-12-7
Molecular Formula C16H18N3NaO3S
Molecular Weight (g/mol) 355.388
MDL Number MFCD00007503
InChI Key YZORUOZKRBVLEG-UHFFFAOYSA-M
Synonym 4-[4-(Diethylamino)phenylazo]benzenesulfonic acid sodium salt
PubChem CID 23676309
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Beilstein 3821641
Formula Weight 355.39
Color Orange
Quantity 100 g
Physical Form Powder
Chemical Name or Material Ethyl Orange sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.