missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate, TRC

Product Code. 17959554
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17959554 1 g 1g
17909554 50 mg 50mg
17958124 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17959554 Supplier Toronto Research Chemicals Supplier No. TRCE9253251G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20 H38 O2
Molecular Weight (g/mol) 310.51
Synonym Ethyl Oleate, (Z)-Octadec-9-enoic Acid Ethyl Ester
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-
Formula Weight 310.2872
Percent Purity >95%
Chemical Name or Material Ethyl Oleate
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.