missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oleate, tech. 70%

Product Code. 11437467
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437467 250 g 250g
11427467 1000 g 1000g
11447467 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437467 Supplier Thermo Scientific Alfa Aesar Supplier No. A15601.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-62-6 | C20H38O2 | 310.522 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-62-6
Molecular Formula C20H38O2
Molecular Weight (g/mol) 310.522
MDL Number MFCD00009579
InChI Key LVGKNOAMLMIIKO-QXMHVHEDSA-N
Synonym ethyl oleate, ethyl cis-9-octadecenoate, oleic acid, ethyl ester, oleic acid ethyl ester, 9-octadecenoic acid z-, ethyl ester, ethyl oleate nf, ethyl z-octadec-9-enoate, ethyl oleate natural, fema no. 2450
PubChem CID 5363269
ChEBI CHEBI:84940
IUPAC Name ethyl (Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)OCC

Specifications

Melting Point -32°C
Density 0.87
Boiling Point 205°C to 208°C
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)7CH=CH(CH2)7CO2CH2CH3
Refractive Index 1.451
Quantity 250 g
Beilstein 1727318
Sensitivity Air Sensitive
Formula Weight 310.53
Percent Purity 70%
Grade Technical
Chemical Name or Material Ethyl oleate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.