missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Oleate-d5, TRC

Product Code. 30384615
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384615 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30384615 Supplier Toronto Research Chemicals Supplier No. TRCE925327100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Oleate-d5
CAS 1217226-00-0
Purity Grade Notes GC
Molecular Formula C20H33D5O2
Quantity 100 mg
Synonym (9Z)-9-Octadecenoic Acid Ethyl Ester-d5, Crodamol EO-d5, Esterol 123-d5, Ethyl (Z)-9-Octadecenoate-d5, Ethyl cis-9-Octadecenoate-d5, Ethyl Oleate-d5, Nikkol EOO-d5, Nofable EO 99-d5, Oleic Acid Ethyl Ester
InChI Formula InChI=1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-/i2D3,4D2
SMILES O=C(OC([2H])([2H])C([2H])([2H])[2H])CCCCCCC/C=C\CCCCCCCC
IUPAC Name 1,1,2,2,2-pentadeuterioethyl (Z)-octadec-9-enoate
Molecular Weight (g/mol) 315.55
Formula Weight 315.3186
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.