missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Octanoate, TRC

Product Code. 30382657
Change view
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382657 50 mL 50mL
30385506 25 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382657 Supplier Toronto Research Chemicals Supplier No. TRCE92531550ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-32-1
Molecular Formula C10 H20 O2
Molecular Weight (g/mol) 172.26
Synonym Octanoic acid-ethyl ester, Octanoic acid, ethyl ester (9CI, ACI), Caprylic acid ethyl ester, Et caprylate, Ethyl caprylate, Ethyl n-octanoate, Ethyl octanoate, NSC 8898
IUPAC Name ethyl octanoate
SMILES CCCCCCCC(=O)OCC

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C10H20O2/c1-3-5-6-7-8-9-10(11)12-4-2/h3-9H2,1-2H3
Formula Weight 172.1463
Chemical Name or Material Caprylic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.