missing translation for 'onlineSavingsMsg'
Learn More

Ethyl octanoate, 99%

Product Code. 11415437
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415437 500 g 500g
11405437 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415437 Supplier Thermo Scientific Alfa Aesar Supplier No. B23450.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-32-1 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-32-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009552
InChI Key YYZUSRORWSJGET-UHFFFAOYSA-N
Synonym ethyl caprylate, octanoic acid, ethyl ester, ethyl n-octanoate, caprylic acid ethyl ester, ethyl octoate, ethyl octylate, octanoic acid ethyl ester, natural, unii-81c5mop582, fema no. 2449
PubChem CID 7799
ChEBI CHEBI:87426
IUPAC Name ethyl octanoate
SMILES CCCCCCCC(=O)OCC

Specifications

Melting Point -47°C
Density 0.872
Boiling Point 206°C to 208°C
Flash Point 79°C (174°F)
Odor Ester-like
Linear Formula CH3(CH2)6CO2CH2CH3
Refractive Index 1.417
Quantity 500 g
Merck Index 14,3778
Formula Weight 172.27
Percent Purity 99%
Chemical Name or Material Ethyl octanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.