missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Nonanoate, TRC

Product Code. 30381077
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381077 10 g -
30379112 5 g 5g
30386818 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381077 Supplier Toronto Research Chemicals Supplier No. TRCE07767510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-29-5
Molecular Formula C11 H22 O2
Molecular Weight (g/mol) 186.29
Synonym Nonanoic acid-ethyl ester, Nonanoic acid, ethyl ester (6CI, 7CI, 8CI, 9CI, ACI), Ethyl n-nonanoate, Ethyl nonanoate, Ethyl nonylate, Ethyl pelargonate, NSC 8901, Pelargonic acid ethyl ester, Wine ether
IUPAC Name ethyl nonanoate
SMILES CCCCCCCCC(=O)OCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H22O2/c1-3-5-6-7-8-9-10-11(12)13-4-2/h3-10H2,1-2H3
Formula Weight 186.162
Chemical Name or Material Nonanoic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.