missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nonanoate, 97%

Product Code. 11451227
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451227 100 mL 100mL
11461227 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451227 Supplier Thermo Scientific Alfa Aesar Supplier No. B24190.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-29-5 | C11H22O2 | 186.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-29-5
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00009570
InChI Key BYEVBITUADOIGY-UHFFFAOYSA-N
Synonym ethyl pelargonate, nonanoic acid, ethyl ester, ethyl nonylate, wine ether, nonanoic acid ethyl ester, natural, ethyl n-nonanoate, unii-ksh683s98j, fema no. 2447, pelargonic acid ethyl ester
PubChem CID 31251
ChEBI CHEBI:87501
IUPAC Name ethyl nonanoate
SMILES CCCCCCCCC(=O)OCC

Specifications

Melting Point -44°C
Density 0.866
Boiling Point 227°C to 229°C
Flash Point 94°C (201°F)
Odor Pleasant
Linear Formula CH3(CH2)7CO2CH2CH3
Refractive Index 1.422
Quantity 100 mL
Beilstein 1759169
Merck Index 14,3838
Formula Weight 186.3
Percent Purity 97%
Chemical Name or Material Ethyl nonanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.