missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Nitroacetate, TRC

Product Code. 30380372
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380372 10 g
30369611 5 g
30382814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380372 Supplier Toronto Research Chemicals Supplier No. TRCE92521510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-35-7
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.1
Synonym 2-Nitroacetic Acid Ethyl Ester, Ethyl 2-Nitroacetate, Ethyl Nitroacetate, NSC 42302, Nitroacetic Acid Ethyl Ester
SMILES CCOC(=O)C[N+](=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H7NO4/c1-2-9-4(6)3-5(7)8/h2-3H2,1H3
Formula Weight 133.0375
Chemical Name or Material Ethyl Nitroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.