missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Nicotinate, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Ethyl Nicotinate |
| CAS | 614-18-6 |
| Molecular Formula | C8 H9 N O2 |
| Quantity | 100 g |
| Synonym | Ethyl Nicotinate, 3-Pyridinecarboxylic acid, ethyl ester, Nicotinic acid, ethyl ester (6CI,8CI), 3-(Ethoxycarbonyl)pyridine, 3-Carbethoxypyridine, Ba 2673, Ethyl 3-pyridinecarboxylate, Ethyl nicotinate, Ignicut, Ignocut, Mucotherm, NSC 8872, Nicaethan, Nikethan, Nikithan, β-Pyridinecarboxylic acid ethyl ester |
| InChI Formula | InChI=1S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3 |
| SMILES | CCOC(=O)c1cccnc1 |
| IUPAC Name | ethyl pyridine-3-carboxylate |
| Molecular Weight (g/mol) | 151.16 |
| Formula Weight | 151.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction