missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Nicotinate, TRC

Product Code. 30371197
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 30371197

Brand: Toronto Research Chemicals TRCE925125500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Nicotinate
CAS 614-18-6
Molecular Formula C8 H9 N O2
Quantity 500 g
Synonym Ethyl Nicotinate, 3-Pyridinecarboxylic acid, ethyl ester, Nicotinic acid, ethyl ester (6CI,8CI), 3-(Ethoxycarbonyl)pyridine, 3-Carbethoxypyridine, Ba 2673, Ethyl 3-pyridinecarboxylate, Ethyl nicotinate, Ignicut, Ignocut, Mucotherm, NSC 8872, Nicaethan, Nikethan, Nikithan, β-Pyridinecarboxylic acid ethyl ester
InChI Formula InChI=1S/C8H9NO2/c1-2-11-8(10)7-4-3-5-9-6-7/h3-6H,2H2,1H3
SMILES CCOC(=O)c1cccnc1
IUPAC Name ethyl pyridine-3-carboxylate
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.